Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

50184-U

Nucleosil® C18 HPLC Column

pore size 120 Å, 5 μm particle size, L × I.D. 12.5 cm × 4.6 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

agency

suitable for USP L1

product line

Nucleosil®

feature

endcapped

manufacturer/tradename

Nucleosil®

extent of labeling

11% Carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

12.5 cm × 4.6 mm

surface area

200 m2/g

matrix

octadecyl

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

120 Å

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Nucleosil is a registered trademark of Macherey-Nagel GmbH & Co. KG

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service