Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

50172-U

Nucleosil® SA (sulphonic acid) HPLC Column

5 μm particle size, L × I.D. 10 cm × 2.1 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

product line

Nucleosil®

feature

endcapped: no

manufacturer/tradename

Nucleosil®

extent of labeling

6.5% Carbon loading

parameter

400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

10 cm × 2.1 mm

surface area

350 m2/g

matrix

sulphonic acid

matrix active group

sulfonic acid phase

particle size

5 μm

pore size

100 Å

separation technique

normal phase

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Nucleosil is a registered trademark of Macherey-Nagel GmbH & Co. KG

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service