Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

501662

Supelco

Carbon Monoxide, Carbon Dioxide, Methane, Ethane, Ethylene and Acetylene

1 % (w/w) each component in nitrogen, aerosol can of 4 L, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

grade

analytical standard

form

gas

packaging

aerosol can of 4 L

manufacturer/tradename

Scott Gas T216-4

concentration

1 % (w/w) each component in nitrogen

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

multi-component solution

Looking for similar products? Visit Product Comparison Guide

General description

Specifications:
Contents
: 4 liters
Pressure: 120 psig
Outlet fitting: Aerosol-type push buttom
Weight: ~100 grams
Dimensions: 2.5 x 8 inches
D.O.T. specifications: 2Q

Application

Carbon monoxide, carbon dioxide, methane, ethane, ethylene and acetylene has been used as a standard analytic gas mixture in the analysis of low molecular weight hydrocarbons using gas chromatography method (GC).[1]

Other Notes

Suitable for qualitative analysis.

Analyte

Description

    Acetylene
    Carbon dioxide (99.8%)
    Carbon monoxide
    Ethane
    Ethylene
    Methane

pictograms

Health hazard

signalword

Danger

Hazard Classifications

Repr. 1A - STOT RE 2 Inhalation

Storage Class

2A - Gases

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of a miniaturised packed gas chromatography column and a SnO 2 gas detector for analysis of low molecular weight hydrocarbons with focus on ethylene detection
Sklorz A, et al.
Sensors and Actuators B, Chemical, 180, 43-49 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service