Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

50102-U

Supelco

Inertsil® ODS-2 HPLC Column

5 μm particle size, L × I.D. 12.5 cm × 4 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
Product 50102-U is not currently sold in your country. Contact Technical Service

feature

endcapped

manufacturer/tradename

Inertsil®

extent of labeling

18.5% Carbon loading

technique(s)

HPLC: suitable

L × I.D.

12.5 cm × 4 mm

matrix

octadecyl

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

150 Å

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Inertsil is a registered trademark of GL Sciences, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service