Key Documents
500852
Eppendorf® CH-500 HPLC column heater system
AC/DC input 240 V AC, Standard European cord
About This Item
UNSPSC Code:
41115854
feature
Standard European cord
packaging
pkg of 1 ea
parameter
ambient to 150°C temp. range
Features and Benefits
This heater/controller essentially combines the features of the CH-30 heater and the TC-50 controller in a single unit. It
includes Eppendorf innovative reversible heating blocks and remote heater on/off capability. Alternatively, the heating
channels for 1/8 in. columns can be used to heat solvent before it enters the injector.
CE/CSA/UL approved.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)