Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

500631

Supelco

ASTM® D2887 Quantitative Calibration Solution

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

analytical standard

agency

ASTM® D2887

packaging

ampule of 1 mL

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

application

ASTM® D2887 quantitative calibration solution has been used as calibration standard to quantitate the alkanes produced during the co-pyrolysis of high density polyethylene to form bio oil, using gas chromatography method coupled with mass spectrometry (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Prepared in carbon disulfide at 0.5 w/% each component unless otherwise noted.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Analyte

Description

    Decane
    Dodecane
    Eicosane
    Heptadecane
    Heptane
    Hexacosane
    Hexadecane 1 % (w/w)
    Hexane
    Nonane
    Octadecane 1 % (w/w)
    Octane
    Pentadecane
    Pentane
    Tetracosane
    Tetradecane
    Undecane
    See All (16)

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1

target_organs

Peripheral nervous system,Central nervous system,Cardio-vascular system,Eyes

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-22.0 °F - closed cup

flash_point_c

-30 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wood/plastic copyrolysis in an auger reactor: Chemical and physical analysis of the products
Bhattacharya P, et al.
Fuel: The Science and Technology of Fuel and Energy, 88, 1251-1260 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service