4S8949
EPA SS TCL Volatiles Mix 1
certified reference material, 2000 μg/mL each component in methanol: water (9:1)
Synonym(s):
EPA TCL Volatiles Mix 1
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
agency
EPA 8260
description
Separate Source
product line
TraceCERT®
CofA
current certificate can be downloaded
feature
standard type calibration
packaging
ampule of 1 × 1 mL
concentration
2000 μg/mL each component in methanol: water (9:1)
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
environmental
format
multi-component solution
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
Other Notes
Legal Information
Analyte
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
target_organs
Eyes,Central nervous system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 2
flash_point_f
49.5 °F - Solvent
flash_point_c
9.7 °C - Solvent
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service