Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

4S8913

Supelco

EPA SS TCL Pesticides Mix

certified reference material, 2000 μg/mL each component in hexane: toluene (1:1)

Synonym(s):

EPA TCL Pesticides Mix

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material

agency

EPA 8081,8270
MISA (Canada) Group 22

description

Separate Source

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 × 1 mL

concentration

2000 μg/mL each component in hexane: toluene (1:1)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Aldrin
    α-BHC
    β-BHC
    Lindane
    δ-BHC
    1,1-Dichloro-2,2-bis(4-chlorophenyl)ethane
    4,4′-DDE
    4,4′-DDT
    Dieldrin
    α-Endosulfan
    β-Endosulfan
    Endosulfan sulfate
    Endrin
    Endrin aldehyde
    Endrin ketone
    Heptachlor
    Heptachlor exo-epoxide
    Methoxychlor
    See All (18)

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT RE 2 Inhalation - STOT SE 3

target_organs

Central nervous system, Nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

40.0 °F

flash_point_c

4.44 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Agata Witczak et al.
Nutrients, 13(1) (2021-01-21)
The aim of the present study was to assess infant safety associated with the occurrence of endocrine-disrupting organochlorine pesticides (OCP) in breast milk. Moreover, the association between pregnant mothers' dietary habits and these compounds levels in breast milk was investigated.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service