Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

4S8905

Supelco

SS TCL Polynuclear Aromatic Hydrocarbons Mix

certified reference material, 2000 μg/mL each component in methylene chloride: benzene (1:1)

Synonym(s):

Polynuclear Aromatic Hydrocarbons Mix

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

agency

EPA 8100,8270
MISA (Canada) Group 19

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 × 1 mL

concentration

2000 μg/mL each component in methylene chloride: benzene (1:1)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

Storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Recommended products

This product is being deleted and is being replaced with product CRM48905

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Acenaphthene
    Acenaphthylene
    Anthracene
    Benz[a]anthracene
    Benzo[b]fluoranthene
    Benzo[k]fluoranthene
    Benzo[ghi]perylene
    Benzo[a]pyrene
    Chrysene
    Dibenz[a,h]anthracene
    Fluoranthene
    Fluorene
    Indeno[1,2,3-cd]pyrene
    Naphthalene
    Phenanthrene
    Pyrene
    See All (16)

signalword

Danger

flash_point_f

12.0 °F

flash_point_c

-11.1 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service