Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

4S8851-U

EPA Purgeable A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

CofA

certificate of analysis is enclosed in each package.

packaging

ampule of 1 mL

concentration

200 μg/mL each component in methanol

Analyte

Description

    Carbon tetrachloride
    Chlorobenzene
    2-Chloroethyl vinyl ether
    Chloroform
    Dibromochloromethane
    1,1-Dichloroethane
    1,1-Dichloroethylene
    Dichloromethane
    1,2-Dichloropropane
    Tetrachloroethylene
    Trichloroethylene
    Trichlorofluoromethane
    See All (12)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service