Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

4S8804

Supelco

EPA VOC Calibration Standards Kit

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

analytical standard

agency

EPA 8260,502.1,502.2,524.1,524.2,8021 (1987),8021A (1990)

description

Separate Source

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

pkg of 1 × 1 kit (4S8804)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Kit Components Only

Product No.
Description

  • EPA Volatile Organic Compounds Mix 1 (4S8775) 1 mL

  • EPA Volatile Organic Compounds Mix 2 (4S8777) 1 mL

  • EPA Volatile Organic Compounds Mix 3 (4S8779) 1 mL

  • EPA Volatile Organic Compounds Mix 4 (4S8786) 1 mL

  • EPA Volatile Organic Compounds Mix 5 (4S8797) 1 mL

  • EPA Volatile Organic Compounds Mix 6 (4S8799) 1.5 mL

related product

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 1A - Flam. Liq. 2 - Muta. 1B - Ozone 1 - STOT RE 2 - STOT SE 1

target_organs

Eyes,Central nervous system, Liver,Kidney

Storage Class

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service