Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

4S8786

Supelco

EPA VOC Mix 4

analytical standard, 2000 μg/mL each component in methanol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

analytical standard

Quality Level

agency

EPA 8260,502.1,502.2,524.1,524.2,8021 (1987),8021A (1990)

feature

standard type calibration

packaging

ampule of 1 × 1 mL (4S8786)

concentration

2000 μg/mL each component in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Bromochloromethane
    Bromoform
    Carbon tetrachloride
    Chloroform
    Dibromomethane
    1,1-Dichloroethane
    2,2-Dichloropropane
    Tetrachloroethylene
    1,1,1-Trichloroethane
    See All (9)

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - Ozone 1 - STOT RE 2 - STOT SE 1

target_organs

Eyes,Central nervous system, Liver,Kidney

wgk_germany

WGK 2

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service