4S7944
1,1,2-Trichloro-1,2,2-trifluoroethane
analytical standard, 2000 μg/mL in methanol
Synonym(s):
1,1,2-Trichloro-1,2,2-trifluoroethane solution
About This Item
Recommended Products
grade
analytical standard
description
Separate Source
CofA
current certificate can be downloaded
feature
standard type calibration
packaging
ampule of 1 × 1 mL
concentration
2000 μg/mL in methanol
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
environmental
format
single component solution
storage temp.
2-8°C
InChI
1S/C2Cl3F3/c3-1(4,6)2(5,7)8
InChI key
AJDIZQLSFPQPEY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - Ozone 1 - STOT SE 1
target_organs
Eyes,Central nervous system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 2
flash_point_f
49.5 °F - Solvent
flash_point_c
9.7 °C - Solvent
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service