Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

4S7426-U

Supelco

EPA CLP Organochlorine Pesticides Mix SS, 1x1mL, Tol:Hex(50-50), 2000ug/mL

certified reference material, 2000 μg/mL each component in hexane: toluene (1:1), Separate Source

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

agency

EPA

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL each component in hexane: toluene (1:1)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

Storage temp.

2-8°C

Recommended products

This product is being deleted and is being replaced by product CRM47426

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Aldrin
    α-BHC
    β-BHC
    Lindane
    δ-BHC
    α-Chlordane
    γ-Chlordane
    1,1-Dichloro-2,2-bis(4-chlorophenyl)ethane
    1,1-Dichloro-2,2-bis(4-chlorophenyl)ethene
    4,4′-DDT
    Dieldrin
    α-Endosulfan
    β-Endosulfan
    Endosulfan sulfate
    Endrin
    Endrin aldehyde
    Endrin ketone
    Heptachlor
    Heptachlor exo-epoxide
    Methoxychlor
    See All (20)

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-14.8 °F

flash_point_c

-26 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jovani Ruiz-Toledo et al.
Insects, 9(2) (2018-05-12)
In this paper, we show the results of investigating the presence of organochlorine pesticides in honey and pollen samples from managed colonies of the honey bee, Apis mellifera L. and of the stingless bee Scaptotrigona mexicana Guérin. Three colonies of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service