Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

49054

Supelco

Methoxychlor

PESTANAL®, analytical standard

Synonym(s):

1,1,1-Trichloro-2,2-bis-(p-methoxyphenyl)ethane, 2,2-Bis(4-methoxyphenyl)-1,1,1-trichloroethane, DMDT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15Cl3O2
CAS Number:
Molecular Weight:
345.65
Beilstein/REAXYS Number:
2057367
EC Number:
MDL number:
UNSPSC Code:
41121813
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

86-88 °C (lit.)

application(s)

agriculture
environmental

format

neat

storage temp.

room temp

SMILES string

COc1ccc(cc1)C(c2ccc(OC)cc2)C(Cl)(Cl)Cl

InChI

1S/C16H15Cl3O2/c1-20-13-7-3-11(4-8-13)15(16(17,18)19)12-5-9-14(21-2)10-6-12/h3-10,15H,1-2H3

InChI key

IAKOZHOLGAGEJT-UHFFFAOYSA-N

Gene Information

mouse ... Esr1(13982)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Pesticide with estrogenic activity.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Koji Satsuma et al.
Applied and environmental microbiology, 78(15), 5313-5319 (2012-05-29)
O-Demethylation of insecticide methoxychlor is well known as a phase I metabolic reaction in various eukaryotic organisms. Regarding prokaryotic organisms, however, no individual species involved in such reaction have been specified and characterized so far. Here we successfully isolated a
Mary Ann Ottinger et al.
Journal of toxicology and environmental health. Part B, Critical reviews, 14(5-7), 413-422 (2011-07-28)
Assessing potential risk associated with exposure to endocrine-disrupting chemicals (EDC) has been difficult due to species specific variation in vulnerability and to both short- and long-term effects produced by EDC. In precocial birds, embryonic exposure to EDC impacts sexual differentiation
A M Cummings
Critical reviews in toxicology, 27(4), 367-379 (1997-07-01)
Estrogens can have a variety of physiological effects, especially on the reproductive system. Chemicals with estrogenic activity that are present in the environment may thus be considered potentially hazardous to development and/or reproduction. Methoxychlor is one such chemical, a chlorinated
T Fukuyama et al.
Toxicology, 289(2-3), 132-140 (2011-08-26)
Immunosuppressive environmental chemicals may increase the potency of allergens and thereby play a role in the development of allergic diseases such as allergic rhinitis, asthma and atopic dermatitis (AD). This study's primary objective was to examine the mechanisms behind the
Hye-Rim Lee et al.
International journal of molecular medicine, 29(5), 883-890 (2012-02-07)
Various endocrine disrupting chemicals (EDCs) are exogenous compounds found in the environment and have the potential to interfere with the endocrine system and hormonal regulation. Among EDCs, bisphenol A (BPA) and 1,1,1-trichloro-2,2-bis(4-methoxyphenol)-ethane [methoxychlor (MXC)] have estrogenic activity resulting in a variety

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service