Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

49049

Supelco

γ-BHC

PESTANAL®, analytical standard

Synonym(s):

Lindane, γ-BHC, 1α,2α,3β,4α,5α,6β-Hexachlorocyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6Cl6
CAS Number:
Molecular Weight:
290.83
Beilstein/REAXYS Number:
1907337
EC Number:
MDL number:
UNSPSC Code:
41121813
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

113-115 °C (lit.)

application(s)

agriculture
environmental

format

neat

storage temp.

2-30°C

SMILES string

Cl[C@H]1[C@H](Cl)[C@H](Cl)[C@@H](Cl)[C@H](Cl)[C@@H]1Cl

InChI

1S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H/t1-,2-,3-,4+,5+,6+

InChI key

JLYXXMFPNIAWKQ-GNIYUCBRSA-N

Gene Information

mouse ... Esr1(13982)
rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

Application

Product 49049 has been discontinued. Product 45548-250MG may be an acceptable replacement.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - STOT RE 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

L A Videla et al.
Free radical biology & medicine, 9(2), 169-179 (1990-01-01)
The development of an oxidative stress condition in the liver by lindane intoxication is discussed as a possible hepatotoxic mechanism of the insecticide. Lindane is metabolized by liver microsomal enzymes to a variety of metabolites, which are susceptible of conjugation
John Vijgen et al.
Environmental science and pollution research international, 18(2), 152-162 (2010-11-26)
Hexachlorocyclohexane (HCH) isomers (α-, β- and γ- (Lindane)) were recently included as new persistent organic pollutants (POPs) in the Stockholm Convention, and therefore, the legacy of HCH and Lindane production became a contemporary topic of global relevance. This article wants
Yuji Nagata et al.
Applied microbiology and biotechnology, 76(4), 741-752 (2007-07-20)
gamma-Hexachlorocyclohexane (gamma-HCH, also called gamma-BHC and lindane) is a halogenated organic insecticide that causes serious environmental problems. The aerobic degradation pathway of gamma-HCH was extensively revealed in bacterial strain Sphingobium japonicum (formerly Sphingomonas paucimobilis) UT26. gamma-HCH is transformed to 2,5-dichlorohydroquinone
Microbial degradation of the pesticide lindane (gamma-hexachlorocyclohexane).
B K Singh et al.
Advances in applied microbiology, 47, 269-298 (2003-07-25)
Kaoruko Mizukawa et al.
Marine pollution bulletin, 70(1-2), 296-302 (2013-03-19)
We analyzed polychlorinated biphenyls (PCBs), dichlorodiphenyl dichloroethane and its metabolites, hexachlorocyclohexanes (HCHs), polycyclic aromatic hydrocarbons (PAHs), and hopanes, in plastic resin pellets collected from nine locations along the Portuguese coast. Concentrations of a sum of 13 PCBs were one order

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service