Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48997-U

Supelco

EPA 8151 Methylated herbicide mix

in acetone (varied), PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

analytical standard

agency

EPA 8151

product line

PESTANAL®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

in acetone (varied)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

General description

EPA 8151 Methylated herbicide mix is useful for identifying and quantifying the chlorinated herbicides on the EPA′s Appendix VIII and Appendix IX lists in solid wastes and groundwater at active hazardous treatment, storage, and disposal sites. The pre-requisite of EPA 8251 method is the use of gas chromatography with a selective detector or gas chromatography/mass spectrometry (GC/MS) or high performance liquid chromatography (HPLC).

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Acifluorfen methyl ester 100 μg/mL
    Bentazon methyl derivative 100 μg/mL
    Chloroamben methyl ester 100 μg/mL
    2,4-D methyl ester 100 μg/mL
    2,4-DB methyl ester 100 μg/mL
    Dacthal dimethyl (DCPA methyl ester) 100 μg/mL
    Dalapon methyl ester 100 μg/mL
    Dicamba methyl ester 100 μg/mL
    3,5-Dichlorobenzoic acid methyl ester 100 μg/mL
    Dichloroprop methyl ester 100 μg/mL
    Dinoseb methyl ether 100 μg/mL
    MCPA methyl ester 10000 μg/mL
    MCPP methyl ester 10000 μg/mL
    4-Nitroanisole 100 μg/mL
    Pentachloroanisole 100 μg/mL
    Picloram methyl ester 100 μg/mL
    2,4,5-T methyl ester 100 μg/mL
    2,4,5-TP methyl ester 100 μg/mL
    See All (18)

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

1.4 °F

flash_point_c

-17 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service