Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48950-U

Supelco

EPA 502 Internal Standard Mix

certified reference material, 2000 μg/mL each component in methanol

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

agency

EPA 502.1,502.2,8021 (1987),8021A (1990)

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type internal

packaging

ampule of 1 mL

concentration

2000 μg/mL each component in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

Storage temp.

2-8°C

General description

EPA 502 Internal Standard Mix is a multi-component solution containing 2-bromo-1-chloropropane and fluorobenzene as certified reference materials (CRMs).
US EPA 500 Series methods are analytical methods for identifying and quantifying organic contaminants such as volatile organic compounds (VOCs), pesticides, synthetic organic compounds (SOCs), and trihalomethane disinfection byproducts (THMs) in raw source water, finished drinking water, and drinking water at all stages of treatment.

application

EPA 502 Internal Standard Mix may be used as a CRM to quantify the disinfection byproducts (DBPs) in chlorine-containing household products by gas chromatography-mass spectrometry (GC-MS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    2-Bromo-1-chloropropane
    Fluorobenzene

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ho-Sang Shin et al.
Chemosphere, 174, 157-164 (2017-02-06)
Seven halogenated volatile organic compounds (HVOCs) and two haloacetic acids were detected and quantified in 15 household products, including sodium hypochlorite, by gas chromatography-mass spectrometry (GC-MS). Chloroform was detected in a range of 0.2-30.2 mg kg-1 in all products, and carbon tetrachloride

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service