Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48937

Supelco

Polystyrene standards, Low molecular weight

analytical standard, pkg of 1 kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

grade

analytical standard

description

individually packaged, 250 mg each

CofA

current certificate can be downloaded

packaging

pkg of 1 kit

technique(s)

gel permeation chromatography (GPC): suitable

format

neat

storage temp.

2-30°C

General description

Low molecular weight polymers with controllable molecular-weight distribution (MWD), microstructure and molecular weight can be prepared by anionic polymerization in solution, in the presence of organolithium compounds.[1] As the molecular weight of the polystyrene increases, physiochemical properties of the impact polystyrene increase, whereas rheological properties decrease.[2]
Molecular weight values are established by size exclusion chromatography, and are nominal values. Actual average molecular weight is within +/- 15% of this value. Certificate of analysis included.

Application

Low molecular weight polystyrene can be used as an adhesive additive to asphalt.[2] Polystyrene standards has been used in the calibration of the molecular weights of aqueous polymer samples obtained from microcalorimetric measurements whose peak molecular weight can be determined using gel permeation chromatography(GPC).[3]
Polystyrene standards, Low molecular weight is used as a calibration standard for the gel permeation chromatographic (GPC) determination of molecular weight measurements of lignin extracted from poplar and pine, in the production of lignin-derived activated carbon (AC) materials.[4]

Kit Components Only

Product No.
Description

  • MW 2500 250 mg

  • MW 5000 250 mg

  • MW 9000 250 mg

  • MW 17,500 250 mg

  • MW 30,000 250 mg

  • MW 50,000 250 mg

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and modification of low-molecular-weight polybutadiene and polystyrene for use as adhesive additive to asphalt.
Pankov IV
Russian Journal of Applied Chemistry, 85 (10), 1627-1630 (2012)
Linking lignin source with structural and electrochemical properties of lignin-derived carbon materials
Li W, et al.
Royal Society of Chemistry Advances, 8(68), 38721-38732 (2018)
Effect of the molecular weight of the polystyrene matrix on the physicomechanical properties of impact polystyrene.
Grmela VP and Farka P
Polym. Chem., 7 (3), 482-484 (1971)
Microcalorimetric detection of lower critical solution temperatures in aqueous polymer solutions
Schild.GH and Tirrell.AD
The Journal of Physical Chemistry, 94(10), 4352-4356 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service