Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48932

Supelco

ASTM® PS-12-60D

in cyclohexane (varied conc.), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352200

grade

analytical standard

agency

ASTM® PS-12-60D

CofA

current certificate can be downloaded

packaging

pkg of 6 × 1 mL

concentration

in cyclohexane (varied conc.)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

single component solution

storage temp.

2-30°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Analyte

Description

    Docosane 800 μg/mL
    Dodecane 200 μg/mL
    Dotriacontane 800 μg/mL
    Eicosane 600 μg/mL
    Hexacontane 200 μg/mL
    Hexacosane 1200 μg/mL
    Hexadecane 400 μg/mL
    Hexatriacontane 600 μg/mL
    Octacosane 1200 μg/mL
    Octadecane 500 μg/mL
    Pentacontane 300 μg/mL
    Tetracontane 500 μg/mL
    Tetracosane 1000 μg/mL
    Tetradecane 300 μg/mL
    Tetratetracontane 400 μg/mL
    Triacontane 1000 μg/mL
    See All (16)

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

-0.4 °F - closed cup

flash_point_c

-18 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service