Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48900-U

Supelco

EPA TCL Base/Neutrals Mix 1

certified reference material, 2000 μg/mL each component in methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

agency

EPA
610

description

Separate Source

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type semivolatile

packaging

ampule of 1 × 1 mL

concentration

2000 μg/mL each component in methylene chloride

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Certified reference material (CRM) standards such as EPA TCL Base/Neutrals Mix 1 are specifically designed for monitoring organic chemicals on the US EPA′s Target Compound List (TCL) in water, sediment, or soil from hazardous waste sites.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Benzyl butyl phthalate
    Bis(2-chloroethoxy)methane
    Bis(2-chloroethyl) ether
    Bis(2-ethylhexyl) phthalate
    4-Bromodiphenyl ether
    4-Chlorodiphenyl ether
    Bis-(2-chloroisopropyl) ether
    Dibutyl phthalate
    Diethyl phthalate
    Dimethyl phthalate
    Di-n-octyl phthalate
    N-Nitrosodimethylamine
    N-Nitrosodiphenylamine
    N-Nitrosodi-n-propylamine
    See All (14)

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service