Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48669

Supelco

Indeno[1,2,3-c,d]pyrene solution

certified reference material, 200 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H12
CAS Number:
Molecular Weight:
276.33
Beilstein/REAXYS Number:
1879312
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

200 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

SMILES string

c1ccc-2c(c1)-c3ccc4ccc5cccc6cc-2c3c4c56

InChI

1S/C22H12/c1-2-7-17-16(6-1)18-11-10-14-9-8-13-4-3-5-15-12-19(17)22(18)21(14)20(13)15/h1-12H

InChI key

SXQBHARYMNFBPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luke Chandler Short et al.
Journal of the American Society for Mass Spectrometry, 18(4), 589-599 (2006-12-26)
The technique of atmospheric pressure photoionization (APPI) has several advantages over electrospray ionization (ESI) and atmospheric pressure chemical ionization (APCI), including efficient ionization of nonpolar or low charge affinity compounds, reduced susceptibility to ion suppression, high sensitivity, and large linear
Nitration of indeno[1,2,3-cd]pyrene and mutagenic activities of related compounds.
M Minabe et al.
Chemical research in toxicology, 2(6), 357-358 (1989-11-01)
Indeno [1,2,3-cd]pyrene.
IARC monographs on the evaluation of the carcinogenic risk of chemicals to humans, 32, 373-378 (1983-12-01)
J E Rice et al.
Carcinogenesis, 11(11), 1971-1974 (1990-11-01)
Indeno[1,2,3-cd]pyrene (IP) is a non-alternant polycyclic aromatic hydrocarbon that has tumor-initiating activity on mouse skin and is carcinogenic in newborn mice and in rat lungs. Previous studies have shown that 8- and 9-hydroxyIP and IP-1,2-diol are major metabolites formed in
J E Rice et al.
Carcinogenesis, 7(10), 1761-1764 (1986-10-01)
Indeno[1,2,3-cd]pyrene is a ubiquitous environmental pollutant which is active as a tumor initiator and complete carcinogen on mouse skin and is carcinogenic in rat lung. The major metabolites of indeno[1,2,3-cd]pyrene as formed in vivo in mouse skin have been identified.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service