Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48667

Supelco

Benzo[ghi]perylene solution

certified reference material, 200 μg/mL in dichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H12
CAS Number:
Molecular Weight:
276.33
Beilstein/REAXYS Number:
1913029
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

200 μg/mL in dichloromethane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

SMILES string

c1cc2ccc3ccc4ccc5cccc6c(c1)c2c3c4c56

InChI

1S/C22H12/c1-3-13-7-9-15-11-12-16-10-8-14-4-2-6-18-17(5-1)19(13)21(15)22(16)20(14)18/h1-12H

InChI key

GYFAGKUZYNFMBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benzo[ghi]perylene.
IARC monographs on the evaluation of the carcinogenic risk of chemicals to humans, 32, 195-204 (1983-12-01)
J R Van Gurp et al.
Cancer research, 43(12 Pt 1), 6031-6038 (1983-12-01)
The development of a selection procedure is described which utilizes the fact that certain polycyclic aromatic hydrocarbons are rendered highly cytotoxic when illuminated with near-ultraviolet light. Twenty polycyclic aromatic hydrocarbons were screened for phototoxicity and toxicity in the absence of
N C Hughes et al.
Carcinogenesis, 14(1), 127-133 (1993-01-01)
Benzo[ghi]perylene (B[ghi]P) is a polycyclic aromatic hydrocarbon (PAH), present in complex combustion products, and evidence for its carcinogenic activity in experimental animals is equivocal, and yet it has demonstrable mutagenic activity in vitro. In order to investigate the possible DNA
Py and BPe solvent polarity scales: effect of temperature on pyrene and benzo[ghi]perylene fluorescence spectra.
R Waris et al.
The Analyst, 113(9), 1465-1467 (1988-09-01)
S H Cherng et al.
Toxicology and applied pharmacology, 170(1), 63-68 (2001-01-06)
Although benzo[g,h,i]perylene (BghiP) has been found to promote the carcinogenesis of benzo[a]pyrene (BaP) in animal models, not much is known about this cocarcinogenic mechanism. In this study, human hepatoma HepG2 cells cotreated with BaP and BghiP were used as a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service