Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48623

Supelco

Dichlorodifluoromethane solution

200 μg/mL in methanol, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CCl2F2
CAS Number:
Molecular Weight:
120.91
EC Number:
UNSPSC Code:
41121813

grade

analytical standard

packaging

ampule of 1 mL

concentration

200 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/CCl2F2/c2-1(3,4)5

InChI key

PXBRQCKWGAHEHS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

51.8 °F - closed cup

flash_point_c

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ya-Fen Wang et al.
Chemosphere, 62(10), 1681-1688 (2005-08-09)
Sulfur hexafluorine compound (SF6), trifluoromethane (CHF3) and diclorodifluoromethane (CCl2F2) are extensively used in the semiconductor industry. They are global warming gases. Most studies have addressed the effective decomposition of fluorine compounds, rather than the toxicity of decomposed by-products. Hence, the
Yukinori Maruo et al.
Dental materials journal, 24(3), 311-314 (2005-11-11)
The purpose of this study was to examine if a modified direct relining method could result in excellent denture adaptation compared to a conventional method. Direct relining was done using heat-polymerized acrylic resin base on a maxillary cast with two
S-K Wang et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 23(43), 435011-435011 (2011-10-15)
Continuous-time core-level photon-stimulated desorption (PSD) spectroscopy was used to investigate the monochromatic soft x-ray photoreactions of CF(2)Cl(2) adsorbed on Si(111)-7 × 7 near the F(1s) edge (681-704 eV). Sequential F(+) PSD spectra were observed as a function of photon exposure
Tian-yang Xiu et al.
Huan jing ke xue= Huanjing kexue, 26(1), 1-6 (2005-04-30)
In recent years, concentration of CFC in the atmosphere has undergone rapid changes especially in association with human activities. For the influence of CFC on climate changes and environment changes, the concentration of atmospheric CFC-11 in Beijing is continuously observed
Shih-Hsiang Lo et al.
Environmental health perspectives, 114(11), 1713-1717 (2006-11-17)
Although there are ample data on the respiratory effects of exposure to fire extinguisher gas, the potential hematologic effects have not been fully documented. We conducted this study to determine the possible etiologic agent(s) for a decrease in red blood

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service