Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48560-U

Supelco

Di-n-octyl phthalate

analytical standard

Synonym(s):

DNOP, Phthalic acid di-n-octyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H38O4
CAS Number:
Molecular Weight:
390.56
Beilstein/REAXYS Number:
1915994
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 5000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

density

0.980 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-30°C

SMILES string

CCCCCCCCOC(=O)c1ccccc1C(=O)OCCCCCCCC

InChI

1S/C24H38O4/c1-3-5-7-9-11-15-19-27-23(25)21-17-13-14-18-22(21)24(26)28-20-16-12-10-8-6-4-2/h13-14,17-18H,3-12,15-16,19-20H2,1-2H3

InChI key

MQIUGAXCHLFZKX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Di-n-octyl phthalate is a straight chain isomer of di(2-ethylhexyl) phthalate (DEHP).[1] It is a component of mixed C6−C10 linear-chain phthalates.[2]

Application

Di-n-octyl phthalate may be used as a reference standard for the determination of the analyte in water[3] and plastic toys[4] by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

hcodes

Hazard Classifications

Aquatic Chronic 4

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

228.2 °F - closed cup

flash_point_c

109.0 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mono-(3-carboxypropyl) phthalate, a metabolite of di-n-octyl phthalate.
Calafat AM, et al.
Journal of Toxicology and Environmental Health. Part A, 69(3), 215-227 (2006)
Gas chromatographic analysis of phthalate esters in plastic toys.
Rastogi SC.
Chromatographia, 47(11-12), 724-724 (1998)
Development of a solid-phase microextraction method for the determination of phthalic acid esters in water.
Prokupkova G, et al.
Analytica Chimica Acta, 457(2), 211-223 (2002)
S Oishi
Archives of toxicology, 64(2), 143-147 (1990-01-01)
Although it is well established that high dose administration of di(2-ethylhexyl) phthalate (DEHP) and its monoester metabolite (MEHP) induces severe testicular atrophy in rats, the mechanisms of this testicular injury is not clear. The present experiment was undertaken to examine
Yujuan Sha et al.
Environmental monitoring and assessment, 124(1-3), 277-287 (2006-12-13)
Samples of water, sediment and suspended particulates were collected from 13 sites in the middle and lower reaches of the Yellow River in China. Phthalic acid esters (PAEs) concentrations in different phases of each sample were determined by Gas Chromatogram

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service