Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48552

Supelco

N-Nitrosodimethylamine

analytical standard

Synonym(s):

Dimethylnitrosamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NNO
CAS Number:
Molecular Weight:
74.08
EC Number:
MDL number:
UNSPSC Code:
41121813
PubChem Substance ID:

grade

analytical standard

Quality Level

agency

EPA TO-7

vapor pressure

5 mmHg ( 20 °C)

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.437 (lit.)

bp

153 °C/774 mmHg (lit.)

density

1.01 g/mL (lit.)

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

CN(C)N=O

InChI

1S/C2H6N2O/c1-4(2)3-5/h1-2H3

InChI key

UMFJAHHVKNCGLG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Nitrosodimethylamine is a potent carcinogen, which can be formed in conditions of wastewater disinfection process, via the reaction between monochloramine and organic nitrogen-containing precursors.[1]

Application

N-Nitrosodimethylamine may be used as an analytical reference standard for the quantification of the analyte in aqueous samples[2][3][4] using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Induces gastric, liver, kidney and lung cancer in mice and rats.

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - Carc. 1B - STOT RE 1

target_organs

Liver

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

141.8 °F - closed cup

flash_point_c

61.0 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N-nitrosodimethylamine (NDMA) removal by reverse osmosis and UV treatment and analysis via LC?MS/MS
Plumlee HM, et al.
Water Research, 42(1-2), 347-355 (2008)
Characterization and fate of N-nitrosodimethylamine precursors in municipal wastewater treatment plants
Mitch.AW and Sedlak.LD
Environmental Science & Technology, 38, 1445-1454 (2004)
Determination of N-Nitrosamines by Gas Chromatography Coupled to Quadrupole?Time-of-Flight Mass Spectrometry in Water Samples
Sieira JB, et al.
Separations, 7(1) (2020)
High-performance liquid chromatography with fluorescence detection for aqueous analysis of nanogram-level N-nitrosodimethylamine
Cha W, et al.
Analytica Chimica Acta, 566(1), 109-116 (2006)
Xiaojie Gan et al.
Water research, 47(3), 1344-1352 (2013-01-05)
The objective of this study was to examine the potential of Magnetic Ion Exchange (MIEX(®)) technology for removing nitrogenous disinfection byproduct (N-DBP) precursors while minimizing carbonaceous DBP (C-DBP) precursors in (i) surface waters, and (ii) effluent impacted waters. Samples were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service