Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48512

Supelco

1,1-Dichloroethane

analytical standard

Synonym(s):

Ethylidene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CHCl2
CAS Number:
Molecular Weight:
98.96
Beilstein/REAXYS Number:
1696901
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

CC(Cl)Cl

InChI

1S/C2H4Cl2/c1-2(3)4/h2H,1H3

InChI key

SCYULBFZEHDVBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Product 48512 is discontinued. Product 36967-1G may be an acceptable replacement.

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

14.0 °F - closed cup

flash_point_c

-10.0 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Howard.HP
Handbook of Environmental Fate and Exposure Data For Organic Chemicals, 2 (1990)
Determining volatile organic compounds in human blood from a large sample population by using purge and trap gas chromatography/mass spectrometry
Ashley DL, et al.
Analytical Chemistry, 64(9), 1021-1029 (1992)
Evaluation of purge-and-trap-high-resolution gas chromatography-mass spectrometry for the determination of 27 volatile organic compounds in marine water at the ng l-1 concentration level
Huybrechts T, et al.
Journal of Chromatography A, 893(2), 367-382 (2000)
Proctor.HN, et al.
Proctor and Hughes' Chemical Hazards of the Workplace (2004)
Gas chromatography-mass spectrometry with headspace for the analysis of volatile organic compounds in waste water
Safarova VI, et al.
Journal of Chromatography. B, Biomedical Applications, 800(1-2), 325-330 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service