About This Item
Recommended Products
grade
analytical standard
product line
PESTANAL®
CofA
current certificate can be downloaded
packaging
ampule of 50 mg
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
agriculture
format
neat
storage temp.
2-30°C
SMILES string
Cl[C@H]1[C@H](Cl)[C@@H](Cl)[C@H](Cl)[C@@H](Cl)[C@@H]1Cl
InChI
1S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H/t1-,2-,3+,4+,5-,6-
InChI key
JLYXXMFPNIAWKQ-CDRYSYESSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Seawater samples by solid-phase extraction (SPE) and gas chromatography coupled to triple quadrupole tandem mass spectrometry (GC-QqQ-MS/MS) equipped with electron impact (EI) source and multiple reaction monitoring (MRM) detection.[2]
- Agricultural products by GC-MS in conjunction with matrix matching and multiple isotopically labeled internal standards.[3]
- Fish samples by QuEChERS (quick, easy, cheap, effective, rugged and safe) extraction, dispersive liquid-liquid microextraction (DLLME) based on solidification of floating organic droplet (SFO) method and GC equipped with electron capture detector (ECD).[4]
- Flos Chrysanthemi by accelerated solvent extraction (ASE) and GC-ECD.[5]
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service