Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48482

Supelco

Polywax® 655

10,000 μg/mL in p-xylene, analytical standard

Synonym(s):

Polywax®

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352200

grade

analytical standard

agency

ASTM® D5307

packaging

pkg of 6 × 1 mL

concentration

10,000 μg/mL in p-xylene

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

single component solution

Looking for similar products? Visit Product Comparison Guide

General description

Polywax® 655 is a blend of polyethylene oligomers with a carbon number range of C10 to C110 in even-number increments.[1]

Application

Polywax® 655 may be used as a calibration standard for the characterization of ultra-heavy oils and bitumens using gas chromatographic technique.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
Polywax is a registered trademark of Baker Hughes, Inc.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

77.0 °F - closed cup

flash_point_c

25 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Compositional analysis of bitumen and bitumen-derived products.
Subramanian M, et al.
Journal of Chromatographic Science, 34(1), 20-26 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service