Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48477

Supelco

Polywax® 655

analytical standard

Synonym(s):

Polywax®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

grade

analytical standard

agency

ASTM® D5307

CofA

current certificate can be downloaded

packaging

ampule of 5000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

neat

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

Polywax® 655, a polyethylene wax, is a high molecular weight hydrocarbon standard for high temperature simulated distillation or gas chromatography analysis.

Application

Polywax® 655 has been used as a standard for the baseline subtraction basically to avoid baseline perturbations, in the simulated distillation of wax samples, using supercritical fluid chromatography and high temperature gas chromatography method of analysis.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
Polywax is a registered trademark of Baker Hughes, Inc.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Simulated distillation of wax samples using supercritical fluid and high temperature gas chromatography
Thomson.S.J and Raynaski.F.R
Journal of Separation Science, 15, 227-234 (1992)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service