Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48395

Supelco

EPA 8010 Halogenated VOC Mix 2

2000 μg/mL each component in methanol, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121813

grade

analytical standard

Quality Level

agency

EPA 8010

feature

standard type calibration

packaging

ampule of 1 mL

concentration

2000 μg/mL each component in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Bromobenzene
    Bromodichloromethane
    Chlorobenzene
    Dibromochloromethane
    1,2-Dichlorobenzene
    1,3-Dichlorobenzene
    1,4-Dichlorobenzene
    1,1-Dichloroethylene
    cis-1,2-Dichloroethylene
    trans-1,2-Dichloroethylene
    Dichloromethane
    cis-1,3-Dichloropropene
    trans-1,3-Dichloropropene
    See All (13)

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

49.5 °F - closed cup - Solvent

flash_point_c

9.7 °C - closed cup - Solvent


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service