Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48338

Supelco

1,2-Dibromo-3-chloropropane solution

certified reference material, 200 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

200 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

InChI

1S/C3H5Br2Cl/c4-1-3(5)2-6/h3H,1-2H2

InChI key

WBEJYOJJBDISQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

51.8 °F - closed cup

flash_point_c

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Albert et al.
Arthritis and rheumatism, 51(4), 656-664 (2004-08-31)
To investigate the possibility that environmental agents contribute to the pathogenesis of Wegener's granulomatosis (WG). We undertook an extensive search for possible environmental exposures by developing a comprehensive questionnaire that was administered by telephone interview to 53 patients with WG
1,2-Dibromo-3-chloropropane.
Report on carcinogens : carcinogen profiles, 12, 134-135 (2011-08-20)
Juraj Lábaj et al.
Nutrition and cancer, 57(2), 209-215 (2007-06-19)
The aim of our investigation was to verify the protective effect of lignin on DNA in rat hepatocytes damaged by 3 different genotoxins attacking DNA in a different manner. Hydrogen peroxide was used for induction of direct single strand breaks
R Sass
International journal of health services : planning, administration, evaluation, 30(3), 491-514 (2000-12-08)
The poisoning of Costa Rican banana workers by multinational corporations' excessive use of pesticides is not a local issue; it is embedded in a dominant ideology expressed by the phenomenon of globalization. This ideology seeps into every aspect of our
Unwelcome exposure.
Tim Rogers
Lancet (London, England), 364(9439), 1025-1026 (2004-09-24)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service