Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48278

Supelco

Carbowax® Amine Test Mix

500 μg/mL each component in methyl tert-butyl ether, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121813

grade

analytical standard

description

Carbowax® Amine

packaging

ampule of 1 mL

concentration

500 μg/mL each component in methyl tert-butyl ether

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

multi-component solution

storage temp.

2-8°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

Carbowax is a registered trademark of Union Carbide

Analyte

Description

    Benzylamine
    Decylamine
    2,4-Dimethylaniline
    2,6-Dimethylaniline
    Eicosane
    Heptadecane
    Hexadecane
    Nonylamine
    Octadecane
    Octylamine
    Pentadecane
    Trihexylamine
    See All (12)

pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

-27.4 °F - closed cup

flash_point_c

-33.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service