Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48234

Supelco

EPA 8010 Surrogate Mix

2000 μg/mL each component in methanol, analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

4 X 1 ML
$194.00

$194.00


Estimated to ship onMay 28, 2025FromMILWAUKEE


Request a Bulk Order

Select a Size

Change View
4 X 1 ML
$194.00

About This Item

UNSPSC Code:
41116107

$194.00


Estimated to ship onMay 28, 2025FromMILWAUKEE


Request a Bulk Order

grade

analytical standard

Quality Level

agency

EPA 8010

feature

standard type surrogate

packaging

ampule of 1 mL

concentration

2000 μg/mL each component in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

General description

EPA 8010 Surrogate Mix can be used for identifying and quantifying the halogenated volatile organics such as bromochloromethane, 1,4-dichlorobutane and 2-bromo-1-chloropropane. In essence, US EPA 8000 Series methods are analytical methods for identifying and quantifying organic compounds on the EPA′s Appendix VIII and Appendix IX lists in solid wastes and groundwater at active hazardous treatment, storage, and disposal sites. The preferred method of quantification is gas chromatography-flame ionization detection (GC-FID), unless otherwise noted.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Bromochloromethane
    2-Bromo-1-chloropropane
    1,4-Dichlorobutane

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - Ozone 1 - STOT SE 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service