48226-U
EPA 8020B Aromatic Volatile Organics Mix 1
certified reference material, 2000 μg/mL each component in methanol
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
agency
EPA 8020
product line
TraceCERT®
CofA
current certificate can be downloaded
feature
standard type calibration
packaging
ampule of 1 mL
concentration
2000 μg/mL each component in methanol
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
environmental
format
multi-component solution
storage temp.
2-8°C
General description
US EPA 8000 Series methods are analytical methods for identifying and quantifying halogenated and aromatic volatile organic compounds on the EPA′s Appendix VIII and Appendix IX lists in solid wastes and groundwater at active hazardous treatment, storage, and disposal sites.
Application
Other Notes
Legal Information
Analyte
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 1A - Flam. Liq. 2 - Muta. 1B - STOT SE 1
target_organs
Eyes,Central nervous system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
51.8 °F
flash_point_c
11 °C
ppe
Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service