48225-U
EPA 8011 EDB/DBCP Mix
certified reference material, 2000 μg/mL each component in methanol
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
agency
EPA 504,8010,8011
product line
TraceCERT®
CofA
current certificate can be downloaded
feature
standard type calibration
packaging
ampule of 1 mL
concentration
2000 μg/mL each component in methanol
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
agriculture
environmental
format
multi-component solution
storage temp.
2-8°C
General description
US EPA 8000 Series methods are analytical methods for identifying and quantifying organic compounds on the EPA′s Appendix VIII and Appendix IX lists in solid wastes and groundwater at active hazardous treatment, storage, and disposal sites.
For ISO/IEC 17025 accredited labs, it is important to show traceability of all results, so for calibration, certified reference materials (CRMs) must be used. Our rapidly expanding TraceCERT® product range includes an increasing number of CRMs for pesticides. These products are manufactured according to ISO/IEC 17025 and ISO Guide 34 double accreditation, which is the highest achievable quality level for reference material producers and which is also referred to as “the gold standard”. The CRM solutions are produced gravimetrically using NIST traceable weights for balance calibration.
Application
Other Notes
Legal Information
Analyte
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Flam. Liq. 2 - Muta. 1B - STOT SE 1
target_organs
Eyes,Central nervous system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
49.5 °F
flash_point_c
9.7 °C
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service