Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48168

Supelco

5-α−Androstane solution

2000 μg/mL in dichloromethane, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H32
CAS Number:
Molecular Weight:
260.46
UNSPSC Code:
12181503

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type internal

packaging

ampule of 1 mL

concentration

2000 μg/mL in dichloromethane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-30°C

InChI

1S/C19H32/c1-18-11-5-7-16(18)15-9-8-14-6-3-4-12-19(14,2)17(15)10-13-18/h14-17H,3-13H2,1-2H3/t14-,15+,16+,17+,18+,19+/m1/s1

InChI key

QZLYKIGBANMMBK-UGCZWRCOSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ivan Cerný et al.
Steroids, 69(3), 161-171 (2004-04-10)
Deuterated analogs of pregnenolone and pregnenolone sulfate with three atoms of deuterium in position 19 were prepared. The synthetic approach was developed on derivatives of dehydroepiandrosterone, where initial intermediates were well characterized, and then applied to the pregnenolone series. Starting
Claudia Cimpoiu et al.
Journal of pharmaceutical and biomedical analysis, 41(2), 633-637 (2006-01-24)
The aim of this paper was the analysis of five androstane isomers by thin-layer chromatography (TLC). The choice of proper mobile phase and the optimization of the mobile phase composition are very important because the chromatographic separation is difficult to
Alexa Greube et al.
Biochemistry, 43(2), 464-472 (2004-01-14)
Seminal plasma of mammalians contains, among others, proteins that are characterized by the fibronectin (Fn) type II module. Our knowledge about the structure and the physiological function of seminal Fn type II proteins mainly originates from studies on PDC-109, the
Verónica Crisóstomo Ayala et al.
Actas urologicas espanolas, 33(8), 895-901 (2009-11-11)
Despite exhibiting histological differences from the human process, canine hormone-induced benign prostatic hyperplasia (BPH) is still the most widely used animal model for evaluating treatment strategies. The aim of this study is to determine the optimal moment for starting a
Neelima Dhingra et al.
Archives of pharmacal research, 34(7), 1055-1063 (2011-08-04)
A number of 17-oxo-5-androsten-3β-yl esters (9a-9f) and 3β-alkoxy-5-androsten-17-ones (11a-11e) were synthesized from commercially available (25R)-5-spirosten-3β-ol (Diosgenin) (4) as starting material. The synthesized compounds were evaluated for their antiproliferative activity against the prostate-specific cancer cell line DU-145, acute toxicity and effect

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service