Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48143

Supelco

EPA TCLP Volatiles Mix

in methanol (varied), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

grade

analytical standard

Quality Level

agency

EPA 1311

feature

standard type calibration

packaging

ampule of 1 mL

concentration

in methanol (varied)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Benzene 1000 μg/mL
    2-Butanone 5000 μg/mL
    Carbon tetrachloride 1000 μg/mL
    Chlorobenzene 1000 μg/mL
    Chloroform 1000 μg/mL
    1,4-Dichlorobenzene 1000 μg/mL
    1,2-Dichloroethane 1000 μg/mL
    1,1-Dichloroethylene 1000 μg/mL
    Tetrachloroethylene 1000 μg/mL
    Trichloroethylene 1000 μg/mL
    Vinyl chloride 1000 μg/mL
    See All (11)

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1A - Flam. Liq. 2 - Muta. 1B - Ozone 1 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

51.8 °F - closed cup

flash_point_c

11 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service