48120-U
EPA TCL Base/Neutrals Mix 2
certified reference material, 2000 μg/mL each component in methylene chloride
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
agency
EPA
610
product line
TraceCERT®
CofA
current certificate can be downloaded
feature
standard type Calibration
packaging
ampule of 1 mL
concentration
2000 μg/mL each component in methylene chloride
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
environmental
format
multi-component solution
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Application
Other Notes
Legal Information
Analyte
signalword
Danger
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service