48100-U
Semivolatile Base-Neutrals Matrix Spike Mix A
1000 μg/mL each component in methanol, analytical standard
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
77101502
NACRES:
NA.24
grade
analytical standard
agency
US EPA/CLP
packaging
ampule of 1 mL
concentration
1000 μg/mL each component in methanol
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
environmental
format
multi-component solution
storage temp.
2-8°C
application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Analyte
Description
Acenaphthene
1,4-Dichlorobenzene
2,4-Dinitrotoluene
N-Nitrosodi-n-propylamine
Pyrene
1,2,4-Trichlorobenzene
signalword
Danger
hcodes
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service