Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48001

Supelco

QTM Volatile Halocarbons Mix

2000 μg/mL each component in methanol, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121813

grade

analytical standard

packaging

ampule of 1 mL

concentration

2000 μg/mL each component in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Bromodichloromethane
    Bromoform
    Carbon tetrachloride
    Chloroform
    1,1-Dichloroethane
    1,2-Dichloroethane
    1,1-Dichloroethylene
    cis-1,2-Dichloroethylene
    trans-1,2-Dichloroethylene
    1,1,2,2-Tetrachloroethane
    Tetrachloroethylene
    1,1,1-Trichloroethane
    Trichloroethylene
    Vinyl chloride
    See All (14)

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1A - Flam. Liq. 2 - Ozone 1 - STOT RE 2 - STOT SE 1

target_organs

Eyes,Central nervous system, Liver,Kidney

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service