Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47992-U

Supelco

EPA TCL Phenols/Benzidines Mix

certified reference material, 2000 μg/mL each component in dichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material

agency

EPA
610

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL each component in dichloromethane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

General description

Certified reference material (CRM) standards such as EPA TCL Phenols/Benzidines Mix are specifically designed for monitoring organic chemicals on the US EPA′s Target Compound List (TCL) in water, sediment, or soil from hazardous waste sites.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Analyte

Description

    Benzidine
    4-Chloro-3-methylphenol
    2-Chlorophenol
    3,3′-Dichlorobenzidine
    2,4-Dichlorophenol
    2,4-Dimethylphenol
    2,4-Dinitrophenol
    2-Methyl-4,6-dinitrophenol
    2-Nitrophenol
    4-Nitrophenol
    Pentachlorophenol
    Phenol
    2,4,6-Trichlorophenol
    See All (13)

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1A - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service