Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47975

Supelco

Pesticide Performance Evaluation Mix-A

in hexane: toluene (99:1) (varied), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121813

grade

analytical standard

Quality Level

packaging

ampule of 1 mL

concentration

in hexane: toluene (99:1) (varied)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

room temp

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    α-BHC
    β-BHC
    Lindane 1 μg/mL
    4,4′-DDT 10 μg/mL
    Decachlorobiphenyl 2 μg/mL
    Endrin 5 μg/mL
    Methoxychlor 25 μg/mL
    2,4,5,6-Tetrachloro-m-xylene 2 μg/mL
    See All (8)

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

target_organs

Central nervous system, Nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-7.6 °F

flash_point_c

-22 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service