Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47970-U

Supelco

EPA HC Purgeable B

certified reference material, 200 μg/mL each component in methanol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

agency

EPA 624

product line

TraceCERT®

feature

standard type calibration

packaging

ampule of 1 mL

concentration

200 μg/mL each component in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Benzene
    Bromodichloromethane
    Bromoform
    1,3-Dichlorobenzene
    1,4-Dichlorobenzene
    1,2-Dichloroethane
    trans-1,2-Dichloroethylene
    cis-1,3-Dichloropropene
    trans-1,3-Dichloropropene
    Ethylbenzene
    1,1,2,2-Tetrachloroethane
    Toluene
    1,1,1-Trichloroethane
    See All (13)

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - STOT SE 1

target_organs

Eyes,Central nervous system

wgk_germany

WGK 2

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service