Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47948

Supelco

Ethyl ether solution

certified reference material, 2000 μg/mL in methanol

Synonym(s):

Diethyl ether solution, Ethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
41121813

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

Storage temp.

2-8°C

InChI

1S/C4H10O/c1-3-5-4-2/h3-4H2,1-2H3

Inchi Key

RTZKZFJDLAIYFH-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F

flash_point_c

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatsuya Yoshino et al.
Organic letters, 14(16), 4290-4292 (2012-08-09)
A highly efficient total synthesis of the 11-membered cyclic aspercyclides A (1) and B (2) has been achieved by chemo- and regioselective intramolecular oxidative C-O bond formation from differently substituted diphenols.
Ai-Bao Xia et al.
The Journal of organic chemistry, 78(3), 1254-1259 (2013-01-05)
The organocatalytic Michael reaction of ketones with γ-monohalonitrodienes catalyzed by chiral prolinethiol ether under solvent-free conditions was developed. The described method represents a novel approach for accessing highly functionalized monohaloalkenes with α, β-stereocenters of up to >99% ee.
Ether-directed ortho-C-H olefination with a palladium(II)/monoprotected amino acid catalyst.
Gang Li et al.
Angewandte Chemie (International ed. in English), 52(4), 1245-1247 (2012-12-15)
Faysal Benaskar et al.
ChemSusChem, 6(2), 353-366 (2012-11-30)
A μ(2)-process in the Ullmann-type C-O coupling of potassium phenolate and 4-chloropyridine was successfully performed in a combined microwave (MW) and microflow process. Selective MW absorption in a micro-fixed-bed reactor (μ-FBR) by using a supported Cu nanocatalyst resulted in an
Gorkem Gunbas et al.
Nature chemistry, 4(12), 1018-1023 (2012-11-24)
Oxatriquinanes are fused, tricyclic oxonium ions that are known to have exceptional stability compared to simple alkyl oxonium salts. C-O bonds in ethers are generally ∼1.43 Å in length, but oxatriquinane has been found to have C-O bond lengths of 1.54 Å.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service