Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47934

Supelco

EPA 524 VOC Mix B

200 μg/mL each component in methanol, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352200

grade

certified reference material

agency

EPA 524.1,524.2

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

200 μg/mL each component in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

for US only, for EU: Cat. No. 32816

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Bromomethane
    Chloroethane
    Chloromethane
    Dichlorodifluoromethane
    Trichlorofluoromethane
    Vinyl chloride

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - Solvent

flash_point_c

9.7 °C - Solvent


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service