Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47908

Supelco

EPA 8270 Organophosphorus Pesticide Mix 2

2000 μg/mL each component in methylene chloride, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816

grade

analytical standard

agency

EPA 8270

feature

standard type calibration

packaging

ampule of 1 mL

concentration

2000 μg/mL each component in methylene chloride

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Dimethoate
    Disulfoton
    Famphur
    Parathion
    Parathion-methyl
    Phorate
    Sulfotep
    Thionazin
    Triethyl thiophosphate
    See All (9)

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service