Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47906

Supelco

EPA Semivolatile Internal Standard Mix, High Concentration

4000 μg/mL each component in methylene chloride: benzene (1:1), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

analytical standard

agency

EPA
610

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

4000 μg/mL each component in methylene chloride: benzene (1:1)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Acenaphthene-d10
    Chrysene-d12
    1,4-Dichlorobenzene-d4
    Naphthalene-d8
    Perylene-d12
    Phenanthrene-d10

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Blood, Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service