Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47896

Supelco

2,4-D solution

100 μg/mL in methanol, PESTANAL®, analytical standard, EPA TCLP

Synonym(s):

2,4-Dichlorophenoxyacetic acid solution, 2,4-D

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
41121813

grade

analytical standard

agency

EPA 1311

product line

PESTANAL®

feature

standard type calibration

packaging

ampule of 1 mL

concentration

100 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C8H6Cl2O3/c9-5-1-2-7(6(10)3-5)13-4-8(11)12/h1-3H,4H2,(H,11,12)

InChI key

OVSKIKFHRZPJSS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - STOT SE 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qian Hou et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 35(5), 675-678 (2012-12-12)
To explore and improve the utilization efficiency of Gentiana macrophylla resources. The germination of Gentiona macrophylla seeds were used as explants to induce calluses; Cultured on various culture media and condition for callus induction and subculture by test; The content
Wafa Tayeb et al.
Toxicology mechanisms and methods, 22(9), 696-704 (2012-08-17)
The present study evaluated the effects of sub-acute exposure to different doses of 2,4-dichlorophenoxyacetic acid (2,4-D) on rat kidney. Forty animals were divided into four equal groups and treated with different doses of 2,4-D: 0, 15, 75 and 150 mg/kg body
Yanhong Tang et al.
Journal of hazardous materials, 241-242, 323-330 (2012-10-16)
A new photocatalyst, Ag nanoparticles (NPs) and reduced graphene oxide (RGO) co-decorated TiO(2) nanotube arrays (NTs) (Ag/RGO-TiO(2) NTs), was designed and facilely produced by combining electrodeposition and photoreduction processes. The structures and properties of the photocatalysts were characterized. The ternary
Erick R Bandala et al.
Environmental technology, 34(1-4), 363-371 (2013-03-28)
Estimation of scaling-up parameters for a site restoration process using a surfactant-enhanced soil washing (SESW) process followed by the application of advanced oxidation processes (Fenton and photo-Fenton) was performed. For the SESW, different parameters were varied and the soil washing
Katherine Coady et al.
Ecotoxicology and environmental safety, 90, 143-150 (2013-01-30)
2,4-Dichlorophenoxyacetic acid (2,4-D) was evaluated in both the Amphibian Metamorphosis Assay (AMA) and the Fish Short Term Reproduction Assay (FSTRA). In the AMA, tadpoles were exposed to mean measured 2,4-D concentrations of 0 (water control), 0.273, 3.24, 38.0 and 113

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service