Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47893

Supelco

JMHW VOC Mix A

1000 μg/mL in methanol, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121813

grade

analytical standard

Quality Level

packaging

ampule of 5 × 1 mL

concentration

1000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Benzene
    Bromodichloromethane
    Bromoform
    Carbon tetrachloride
    Chloroform
    Dibromochloromethane
    1,2-Dichloroethane
    1,1-Dichloroethylene
    cis-1,2-Dichloroethylene
    Dichloromethane
    cis-1,3-Dichloropropene
    trans-1,3-Dichloropropene
    Tetrachloroethylene
    1,1,1-Trichloroethane
    1,1,2-Trichloroethane
    Trichloroethylene
    See All (16)

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 1A - Flam. Liq. 2 - Muta. 1B - Ozone 1 - STOT SE 1

target_organs

Eyes,Central nervous system

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service